Learn More
Thermo Scientific™ ProSwift? RP-4H Capillary Monolithic HPLC Columns
For fast, high-resolution LC-MS separations in protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific? ProSwift? RP-4H Capillary Monolithic HPLC Columns.
Manufacturer:  Thermo Scientific™ 164922
 View more versions of this product
Catalog No. 16-492-2
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
For fast, high-resolution LC-MS separations in protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ ProSwift™ Monolithic HPLC Columns. Based on a ethylvinylbenzene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.
Each monolith is a single cylindrical polymer rod containing an uninterrupted, interconnected network of flow-through channels of a specific pore size. These large channels and the monolith's nonporous surfaces provide fast mass transfer for the high-resolution and fast protein separations. The monolith morphology results in low operating backpressure, allowing the use of higher linear velocities with minimal loss of resolution.
Advantages of High Resolution at Exceptionally High Flow Rates
  • Highest operational flow rates available
  • High throughput and improved productivity
  • Excellent stability over a wide pH range of 1 to 14
  • Outstanding reproducibility and ruggedness
  • High loading capacity
Fast, Sensitive Protein Separations
  • High speed protein separations
  • High sensitivity for LC-MS
  • High column-to-column reproducibility
  • Wide range of column I.D. and lengths available
  • Superior lifetime
Easy Column Installation
  • Use Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
Preconcentrate and Desalt Samples   
  • Precolumns are available to preconcentrate and desalt samples.
  • You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds.
  • Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases.
Description & Specifications
Specifications
Phenyl
Reversed Phase
500 to 3000nL/min
250mm
4950psi
Monolith
Most common Organic Solvents
Analytical Column
Analytical Column
0.1mm
LC/MS
Poly(ethylvinylbenzene-co-divinylbenzene)
Monolith Polymer
1 to 14
70°C
Phenyl