Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protamine Sulfate, Affymetrix/USB™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
9009-65-8
0.00
MFCD00132091
9009-65-8
0.00
MFCD00132091
Packaging Plastic bottle
Quantity 25g
Physical Form Powder
Chemical Name or Material Protamine Sulfate