Learn More
Proteasome 20S alpha 5 Mouse anti-Human, Xenopus, DyLight™ 405, Clone: AH1.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP276657V
Catalog No. NBP276657V
Add to cart
Description
Description
Proteasome 20S alpha 5 Monoclonal antibody specifically detects Proteasome 20S alpha 5 in Human,Xenopus samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Proteasome 20S alpha 5
AH1.1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Xenopus
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight™ 405
50 mM sodium borate with 0.05% sodium azide
PSMA5
Sub-unit 5a
0.1 mL
Primary
5686