Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome beta 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Proteasome beta 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Proteasome beta 1 Polyclonal specifically detects Proteasome beta 1 in Human samples. It is validated for Western Blot.Specifications
Proteasome beta 1 | |
Polyclonal | |
Rabbit | |
P20618 | |
5689 | |
Synthetic peptides corresponding to PSMB1(proteasome (prosome, macropain) subunit, beta type, 1) The peptide sequence was selected from the middle region of PSMB1 (NP_002784). Peptide sequence NNKAMTTGAIAAMLSTILYSRRFFPYYVYNIIGGLDEEGKGAVYSFDPVG. | |
Primary | |
26 kDa |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.4.25.1, FLJ25321, HC5, KIAA1838, Macropain subunit C5, Multicatalytic endopeptidase complex subunit C5, PMSB1, proteasome (prosome, macropain) subunit, beta type, 1, proteasome beta 1 subunit, Proteasome component C5, Proteasome gamma chain, proteasome subunit beta type-1, proteasome subunit HC5, PSC5 | |
PSMB1 | |
IgG | |
This product is specific to Subunit or Isoform: beta type-1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title