Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome subunit beta type 4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15458620UL

 View more versions of this product

Catalog No. NBP15458620


Add to cart

Description

Description

Proteasome subunit beta type 4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Proteasome subunit beta type 4
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PSMB4
Synthetic peptides corresponding to PSMB4(proteasome (prosome, macropain) subunit, beta type, 4) The peptide sequence was selected from the middle region of PSMB4 (NP_002787). Peptide sequence SRRSKMNPLWNTMVIGGYADGESFLGYVDMLGVAYEAPSLATGYGAYLAQ.
29 kDa
20ul
Stem Cell Markers
Primary
5692
Human
Western Blot
Western Blot 0.2-1 ug/ml
P28070
EC 3.4.25.1, HN3, hsBPROS26, HsN3, Macropain beta chain, Multicatalytic endopeptidase complex beta chain, PROS26PROS-26, proteasome (prosome, macropain) subunit, beta type, 4, Proteasome beta chain, Proteasome chain 3, proteasome subunit beta type-4, proteasome subunit HsN3, proteasome subunit, beta type, 4,26 kDa prosomal protein
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This product is specific to Subunit or Isoform: beta type-4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit