Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DenLine Protection Plus+ II Unisex Long-Length Lab Coats, Color: Black
Provides 360 deg. employee protection and comfort needs with high breathable front and back fabric.
Manufacturer: DenLine DL270BJBLK5XL
Description
- Fluid Resistance: Incidental Splash
- Stain resistant to all aqueous liquids like blood, bleach, select reagents
- Lint free fabric; Clean Room Level ISO 5 (Class 100)
- Three pockets
- Double stitched
- Bar Tack reinforced
- Side vents
- Tapered sleeves prevents catching on things when reaching
- Spun woven, low pill, “Quick Dry” cuffs
- Heavy duty stainless steel snaps (lifetime guaranteed)
- For bloodborne applications
Specifications
- Length: 104cm (41 in.)
- Hydrostatic Pressure (AATCC 127): >200mm
- Air Permeability (ATSM D 737): 40 cu. ft./min.
- Breathability: 50% Poly/50% Cotton
- Static Resistance: Surface Resistivity 109 (Test Method 76)
Wash Life
- 200+ Wash Life - Commercial Wash Tested through 225 Hot Water Wash Cycles, Meets CDC HICPAC Recommendations for Processing Medical Linen.
Specifications
99% polyester and 1% carbon | |
Long-Length | |
Black | |
For bloodborne applications | |
3 | |
62 to 64 in. | |
32 to 34 in. | |
158 to 163 cm | |
104 cm | |
Unisex |
No | |
Snaps | |
Knit | |
Polycotton blend | |
5X-Large | |
58 to 61.5 in. | |
147 to 156 cm | |
Tapered | |
41 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title