Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Protection Plus+™ II Unisex Long-Length Lab Coats, Color: Ceil Blue

Provides 360 deg. employee protection and comfort needs with high breathable front and back fabric.

$97.70 - $102.90

Specifications

Closure Type Snaps
Construction 99% polyester and 1% carbon
Cuff Style Knit
Disposable No
For Use With (Application) For bloodborne applications
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
22-412-357
View Documents
DenLine™
DL270BJCEI4XL
4X-Large
Each for $97.70
Add to cart
 
22-412-358
View Documents
DenLine™
DL270BJCEI5XL
5X-Large
Each for $102.90
Add to cart
 
Description

Description

  • Fluid Resistance: Incidental Splash
  • Stain resistant to all aqueous liquids like blood, bleach, select reagents
  • Lint free fabric; Clean Room Level ISO 5 (Class 100)
  • Three pockets
  • Double stitched
  • Bar Tack reinforced
  • Side vents
  • Tapered sleeves prevents catching on things when reaching
  • Spun woven, low pill, “Quick Dry” cuffs
  • Heavy duty stainless steel snaps (lifetime guaranteed)
  • For bloodborne applications

Specifications

  • Length: 104cm (41 in.)
  • Hydrostatic Pressure (AATCC 127): >200mm
  • Air Permeability (ATSM D 737): 40 cu. ft./min.
  • Breathability: 50% Poly/50% Cotton
  • Static Resistance: Surface Resistivity 109 (Test Method 76)

    Wash Life

    • 200+ Wash Life - Commercial Wash Tested through 225 Hot Water Wash Cycles, Meets CDC HICPAC Recommendations for Processing Medical Linen.
Specifications

Specifications

Snaps
Knit
For bloodborne applications
104 cm
Polycotton blend
Incidental splash, stain resistant to all aqueous liquids like blood, bleach, select reagents
Long-Length
99% polyester and 1% carbon
No
Unisex
41 in.
Long-Length Lab Coat
Tapered
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit