Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Protection Plus+™ II Unisex Long-Length Lab Coats, Color: Royal
SDP

Click to view available options
Size:
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides 360 deg. employee protection and comfort needs with high breathable front and back fabric.

  • Fluid Resistance: Incidental Splash
  • Stain resistant to all aqueous liquids like blood, bleach, select reagents
  • Lint free fabric; Clean Room Level ISO 5 (Class 100)
  • Three pockets
  • Double stitched
  • Bar Tack reinforced
  • Side vents
  • Tapered sleeves prevents catching on things when reaching
  • Spun woven, low pill, “Quick Dry” cuffs
  • Heavy duty stainless steel snaps (lifetime guaranteed)
  • For bloodborne applications

Specifications

  • Length: 104cm (41 in.)
  • Hydrostatic Pressure (AATCC 127): >200mm
  • Air Permeability (ATSM D 737): 40 cu. ft./min.
  • Breathability: 50% Poly/50% Cotton
  • Static Resistance: Surface Resistivity 109 (Test Method 76)

    Wash Life

    • 200+ Wash Life - Commercial Wash Tested through 225 Hot Water Wash Cycles, Meets CDC HICPAC Recommendations for Processing Medical Linen.
Type Long-Length Lab Coat
Size 4X-Large
Gender Unisex
Material Polycotton blend
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable No
Resistance Incidental splash, stain resistant to all aqueous liquids like blood, bleach, select reagents
Style Long-Length
No. of Pockets 3
Sleeve Style Tapered
Size (English) Chest 58 to 60 in.
Size (English) Hip 54 to 57.5 in.
Size (English) Waist 28 to 30 in.
Size (Metric) Chest 147 to 152 cm
Size (Metric) Hip 137 to 146 cm
For Use With (Application) For bloodborne applications
Length (English) 41 in.
Length (Metric) 104 cm
Construction 99% polyester and 1% carbon
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit