Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DenLine™ Protection Plus+™ II Unisex Long-Length Lab Coats, Color: Royal
Provides 360 deg. employee protection and comfort needs with high breathable front and back fabric.
$93.00 - $98.00
Specifications
Closure Type | Snaps |
---|---|
Construction | 99% polyester and 1% carbon |
Cuff Style | Knit |
Disposable | No |
For Use With (Application) | For bloodborne applications |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
22-412-355
|
DenLine™
DL270BJROY4XL |
4X-Large |
Each for $93.00
|
|
22-412-356
|
DenLine™
DL270BJROY5XL |
5X-Large |
Each for $98.00
|
|
Description
- Fluid Resistance: Incidental Splash
- Stain resistant to all aqueous liquids like blood, bleach, select reagents
- Lint free fabric; Clean Room Level ISO 5 (Class 100)
- Three pockets
- Double stitched
- Bar Tack reinforced
- Side vents
- Tapered sleeves prevents catching on things when reaching
- Spun woven, low pill, “Quick Dry” cuffs
- Heavy duty stainless steel snaps (lifetime guaranteed)
- For bloodborne applications
Specifications
- Length: 104cm (41 in.)
- Hydrostatic Pressure (AATCC 127): >200mm
- Air Permeability (ATSM D 737): 40 cu. ft./min.
- Breathability: 50% Poly/50% Cotton
- Static Resistance: Surface Resistivity 109 (Test Method 76)
Wash Life
- 200+ Wash Life - Commercial Wash Tested through 225 Hot Water Wash Cycles, Meets CDC HICPAC Recommendations for Processing Medical Linen.
Specifications
Snaps | |
Knit | |
For bloodborne applications | |
104 cm | |
Polycotton blend | |
Incidental splash, stain resistant to all aqueous liquids like blood, bleach, select reagents | |
Long-Length |
99% polyester and 1% carbon | |
No | |
Unisex | |
41 in. | |
Long-Length Lab Coat | |
Tapered |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title