Learn More
Labconco™ Fume Hood Accessory, SpillStopper™ Work Surface
SpillStopper Work Surface is designed for protecting your Labconco Protector™ Echo Filtered, Airo Filtered or XL Fume Hood from accidental spills.
Manufacturer:  Labconco™ 9501811
 View more versions of this product
Catalog No. 10-369-303
  Check Availability
Add to cart
Specifications
Description
  • Molded from special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to interior liner of Protector Echo, Airo and XL hoods
  • Pre-drilled 5.8cm (1.5 in.) diameter holes for venting
  • Pre-drilled 6.4cm (2.5 in.) for plumbing pass-through
  • Front edge has large radius to aerodynamically direct airflow into hood
  • May be ordered with a pre-cut 15.2 x 7.6cm (6 x 3 in.) oval cupsink cutout(s) in left rear, right rear, left side, or right side
  • Cupsink sold separately
Description & Specifications
Specifications
36 in.
91.4cm
8 ft. Protector Echo Filtered, Airo Filtered or XL Fume Hood
3.2cm
113kg
243.8cm
Cutout: Left and right rear
0.4 in.
1.0cm
1.25 in.
250 lb.
96 in.
Corrosion-resistant epoxy resin