Learn More
Labconco™ Protector™ Echo Filtered, Airo Filtered or XL Fume Hood Accessory, SpillStopper™ Work Surfaces
SDPSCI_ED
The Labconco™ Protector™ SpillStopper™ Work Surface is a durable fume hood accessory that helps keep work spaces safe and clean.
Supplier:  Labconco™ 9501402/EMD
Catalog No. S28285
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Molded from special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to interior liner of Protector Echo, Airo and XL hoods
  • Pre-drilled 5.8cm (1.5 in.) diameter holes for venting
  • Pre-drilled 6.4cm (2.5 in.) for plumbing pass-through
  • Front edge has large radius to aerodynamically direct airflow into hood
  • May be ordered with a pre-cut 15.2 x 7.6cm (6 x 3 in.) oval cupsink cutout(s) in left rear, right rear, left side, or right side
  • Cupsink sold separately
Specifications
Specifications
Protector Airo Laboratory Hoods, Protector Echo Laboratory Hoods, Protector XL Laboratory Hoods
36 in.
Right Side Cupsink Cutout
1.25 in.
48 in.
Product Suggestions
Product Suggestions