Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Echo™ Floor-Mounted Filtered Fume Hoods: 4 ft. Width

Fully-featured, energy-efficient fume hood provides an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.

$35010.00 - $37830.00

Specifications

Certifications/Compliance UL 61010-1; CAN/CSA C22.2 No. 61010.1; ANSI Z9.5; SEFA 9, DH III; Modified ASHRAE 110; ASTM E84-09 C; AFNOR NF-X 15-211; SEFA 8, Cabinet Surface Finish Tests
Color Glacier White
For Use With General Chemistry; Large Equipment
Height Overall (English) 102.2 in.
Hertz 50/60 Hz
View More Specs

Alert:

The Lifetime Payback Questionnaire (LPQ) form is REQUIRED for all Protector Echo Floor-Mounted Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.

Requires: Filters, Remote Blower, Ductwork


Products 40
Catalog Number Mfr. No. Depth (English) Exterior Includes Description Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Exterior Includes Description Electrical Requirements Price Quantity & Availability  
13-118-640
View Documents
Labconco™
183417200
37.7 in. 0 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $35,010.00
Add to cart
 
13-118-641
View Documents
Labconco™
183417201
37.7 in. 2 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $35,910.00
Add to cart
 
13-118-642
View Documents
Labconco™
183417202
37.7 in. 2 fixtures, 1 electrical receptacle Carbon 115 V, 50/60 Hz, 10 A Each for $36,035.00
Add to cart
 
13-118-643
View Documents
Labconco™
183417220
37.7 in. 0 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $35,010.00
Add to cart
 
13-118-644
View Documents
Labconco™
183417221
37.7 in. 2 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $35,910.00
Add to cart
 
13-118-645
View Documents
Labconco™
183417400
37.7 in. 0 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $36,510.00
Add to cart
 
13-118-646
View Documents
Labconco™
183417401
37.7 in. 2 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $37,410.00
Add to cart
 
13-118-647
View Documents
Labconco™
183417402
37.7 in. 2 fixtures, 1 electrical receptacle Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $37,535.00
Add to cart
 
13-118-648
View Documents
Labconco™
183417420
37.7 in. 0 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $36,510.00
Add to cart
 
13-118-649
View Documents
Labconco™
183417421
37.7 in. 2 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $37,410.00
Add to cart
 
13-118-650
View Documents
Labconco™
183417600
37.7 in. 0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $36,510.00
Add to cart
 
13-118-651
View Documents
Labconco™
183417601
37.7 in. 2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $37,410.00
Add to cart
 
13-118-652
View Documents
Labconco™
183417602
37.7 in. 2 fixtures, 1 electrical receptacle Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $37,535.00
Add to cart
 
13-118-653
View Documents
Labconco™
183417620
37.7 in. 0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $36,510.00
Add to cart
 
13-118-654
View Documents
Labconco™
183417621
37.7 in. 2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $37,410.00
Add to cart
 
13-118-655
View Documents
Labconco™
183417800
37.7 in. 0 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $35,010.00
Add to cart
 
13-118-656
View Documents
Labconco™
183417801
37.7 in. 2 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $35,910.00
Add to cart
 
13-118-657
View Documents
Labconco™
183417802
37.7 in. 2 fixtures, 1 electrical receptacle HEPA only 115 V, 50/60 Hz, 10 A Each for $36,035.00
Add to cart
 
13-118-658
View Documents
Labconco™
183417820
37.7 in. 0 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $35,010.00
Add to cart
 
13-118-659
View Documents
Labconco™
183417821
37.7 in. 2 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $35,910.00
Add to cart
 
13-118-660
View Documents
Labconco™
183427200
43.7 in. 0 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $35,305.00
Add to cart
 
13-118-661
View Documents
Labconco™
183427201
43.7 in. 2 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $36,205.00
Add to cart
 
13-118-662
View Documents
Labconco™
183427202
43.7 in. 2 fixtures, 1 electrical receptacle Carbon 115 V, 50/60 Hz, 10 A Each for $36,330.00
Add to cart
 
13-118-663
View Documents
Labconco™
183427220
43.7 in. 0 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $35,305.00
Add to cart
 
13-118-664
View Documents
Labconco™
183427221
43.7 in. 2 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $36,205.00
Add to cart
 
13-118-665
View Documents
Labconco™
183427400
43.7 in. 0 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $36,805.00
Add to cart
 
13-118-666
View Documents
Labconco™
183427401
43.7 in. 2 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $37,705.00
Add to cart
 
13-118-667
View Documents
Labconco™
183427402
43.7 in. 2 fixtures, 1 electrical receptacle Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $37,830.00
Add to cart
 
13-118-668
View Documents
Labconco™
183427420
43.7 in. 0 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $36,805.00
Add to cart
 
13-118-669
View Documents
Labconco™
183427421
43.7 in. 2 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $37,705.00
Add to cart
 
13-118-670
View Documents
Labconco™
183427600
43.7 in. 0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $36,805.00
Add to cart
 
13-118-671
View Documents
Labconco™
183427601
43.7 in. 2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $37,705.00
Add to cart
 
13-118-672
View Documents
Labconco™
183427602
43.7 in. 2 fixtures, 1 electrical receptacle Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $37,830.00
Add to cart
 
13-118-673
View Documents
Labconco™
183427620
43.7 in. 0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $36,805.00
Add to cart
 
13-118-674
View Documents
Labconco™
183427621
43.7 in. 2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $37,705.00
Add to cart
 
13-118-675
View Documents
Labconco™
183427800
43.7 in. 0 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $35,305.00
Add to cart
 
13-118-676
View Documents
Labconco™
183427801
43.7 in. 2 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $36,205.00
Add to cart
 
13-118-677
View Documents
Labconco™
183427802
43.7 in. 2 fixtures, 1 electrical receptacle HEPA only 115 V, 50/60 Hz, 10 A Each for $36,330.00
Add to cart
 
13-118-678
View Documents
Labconco™
183427820
43.7 in. 0 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $35,305.00
Add to cart
 
13-118-679
View Documents
Labconco™
183427821
43.7 in. 2 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $36,205.00
Add to cart
 
Description

Description

The Protector™ Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab's Neutrodine™ Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured floor-mount fume hood that requires NO DUCTING for easy installation! Neutrodine Unisorb filters are compatible with over 700 chemicals.

Features (All Models)

  • Eco-Foil™ air foil with aerodynamic Clean-Sweep™ airflow openings
  • Cord-Keeper™ Slots on left and right side of air foil
  • Single piece, tempered safety glass vertical-rising sashes with cable pulley, sash weight and powder-coated aluminum sash handle and tracks with Clean-Sweep™ airflow openings. Upper sash stop at 16 in. sash open height. Upper sash opens to 26 in. high for loading chemicals. Lower sash opens up from floor to 55.2 in. for transporting carts, drums and apparatus
  • Removable front and side panels, and front and interior service access panels for access to plumbing and electrical wiring
  • eGuard2 software communication
  • Smart control module with wifi, Bluetooth and ethernet connectivity
  • Factory-installed command module, fan button and light button
  • Rear baffle
  • By-pass airflow design
  • Glacier white powder-coated steel exterior
  • Chemical-resistant, fiberglass-reinforced, composite panel liner
  • Pre-wired vapor-proof LED lights for up to 63 foot candles rated for 50,000 hours: 2 lights per 4 ft. model, 3 lights per 5 ft. model, 4 lights per 6 ft. model, and 5 lights per 8 ft. model
  • Factory-prepared for up to two electrical duplex receptacles and six service fixtures
  • Roughing prefilter: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model
  • Built-in exhaust fans: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model, to maintain 60 fpm face velocity with only an upper sash opening height of 16 in.
  • Sound pressure of <60 dB(A) with sash at operating height and <48 dB(A) with sash closed
  • Sensor package for primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position, and temperature
  • Audible/visual alarms for breakthrough detection, temperature ≥40°C (104°F), fan failure and high sash position opening. At ≥60°C (140°F), all fans stop
  • Power cord with plug

Fixtured Models May Feature

  • Two pre-plumbed service fixtures with forged brass valves, lower right side with brass tubing for gas and lower left side with copper tubing for cold water. Components for converting either or both fixtures to air and vacuum are provided. Inlet tubing is not provided
  • One pre-wired GFCI electrical duplex receptacle on lower right side

Required Accessories (Not Included)

  • Neutrodine™ Unisorb Filters (except HEPA only models which require HEPA filters). Number of filters required is determined by the width of the hood

Options Include

  • HEPA filter
  • Region-specific electrical plug configurations
  • Service fixtures

Standards Conformance and Regulations

  • SEFA 9, DHIII
  • SEFA 8, Cabinet Surface Finish Tests
  • MODIFIED ASHRAE 110
  • ANSI Z9.5-2011
  • AFNOR NF-X 15-211
  • ASTM E84-09C
  • UL Standard 61010-1
  • CAN/CSA C22.2 No. 61010-1
Specifications

Specifications

UL 61010-1; CAN/CSA C22.2 No. 61010.1; ANSI Z9.5; SEFA 9, DH III; Modified ASHRAE 110; ASTM E84-09 C; AFNOR NF-X 15-211; SEFA 8, Cabinet Surface Finish Tests
General Chemistry; Large Equipment
50/60 Hz
Chemical-resistant, fiberglass-reinforced, composite panel
Powder-Coated Steel
52 in.
4 ft.
Glacier White
102.2 in.
LED rated for 50,000 hours
Floor-Mounted
Protector™ Echo™ Floor-Mounted
Filtered Fume Hood

Filters required (sold separately)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit