Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Echo™ Floor-Mounted Filtered Fume Hoods: 5 ft. Width

Fully-featured, energy-efficient fume hood provides an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.

$37355.00 - $39880.00

Specifications

Certifications/Compliance UL 61010-1; CAN/CSA C22.2 No. 61010.1; ANSI Z9.5; SEFA 9, DH III; Modified ASHRAE 110; ASTM E84-09 C; AFNOR NF-X 15-211; SEFA 8, Cabinet Surface Finish Tests
Color Glacier White
For Use With General Chemistry; Large Equipment
Depth (English) Exterior 37.7 in.
Height Overall (English) 102.2 in.
View More Specs

Alert:

The Lifetime Payback Questionnaire (LPQ) form is REQUIRED for all Protector Echo Floor-Mounted Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.

Requires: Filters, Remote Blower, Ductwork


Products 20
Catalog Number Mfr. No. Includes Description Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Includes Description Electrical Requirements Price Quantity & Availability  
13-118-700
View Documents
Labconco™
183517200
0 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $37,355.00
Add to cart
 
13-118-701
View Documents
Labconco™
183517201
2 fixtures, 0 electrical receptacles Carbon 115 V, 50/60 Hz, 10 A Each for $38,255.00
Add to cart
 
13-118-702
View Documents
Labconco™
183517202
2 fixtures, 1 electrical receptacle Carbon 115 V, 50/60 Hz, 10 A Each for $38,380.00
Add to cart
 
13-118-703
View Documents
Labconco™
183517220
0 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $37,355.00
Add to cart
 
13-118-704
View Documents
Labconco™
183517221
2 fixtures, 0 electrical receptacles Carbon 230 V, 50/60 Hz, 5 A Each for $38,255.00
Add to cart
 
13-118-705
View Documents
Labconco™
183517400
0 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $38,855.00
Add to cart
 
13-118-706
View Documents
Labconco™
183517401
2 fixtures, 0 electrical receptacles Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $39,755.00
Add to cart
 
13-118-707
View Documents
Labconco™
183517402
2 fixtures, 1 electrical receptacle Carbon with acid sensor 115 V, 50/60 Hz, 10 A Each for $39,880.00
Add to cart
 
13-118-708
View Documents
Labconco™
183517420
0 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $38,855.00
Add to cart
 
13-118-709
View Documents
Labconco™
183517421
2 fixtures, 0 electrical receptacles Carbon with acid sensor 230 V, 50/60 Hz, 5 A Each for $39,755.00
Add to cart
 
13-118-710
View Documents
Labconco™
183517600
0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $38,855.00
Add to cart
 
13-118-711
View Documents
Labconco™
183517601
2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $39,755.00
Add to cart
 
13-118-712
View Documents
Labconco™
183517602
2 fixtures, 1 electrical receptacle Carbon with formaldehyde sensor 115 V, 50/60 Hz, 10 A Each for $39,880.00
Add to cart
 
13-118-713
View Documents
Labconco™
183517620
0 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $38,855.00
Add to cart
 
13-118-714
View Documents
Labconco™
183517621
2 fixtures, 0 electrical receptacles Carbon with formaldehyde sensor 230 V, 50/60 Hz, 5 A Each for $39,755.00
Add to cart
 
13-118-715
View Documents
Labconco™
183517800
0 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $37,355.00
Add to cart
 
13-118-716
View Documents
Labconco™
183517801
2 fixtures, 0 electrical receptacles HEPA only 115 V, 50/60 Hz, 10 A Each for $38,255.00
Add to cart
 
13-118-717
View Documents
Labconco™
183517802
2 fixtures, 1 electrical receptacle HEPA only 115 V, 50/60 Hz, 10 A Each for $38,380.00
Add to cart
 
13-118-718
View Documents
Labconco™
183517820
0 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $37,355.00
Add to cart
 
13-118-719
View Documents
Labconco™
183517821
2 fixtures, 0 electrical receptacles HEPA only 230 V, 50/60 Hz, 5 A Each for $38,255.00
Add to cart
 
Description

Description

The Protector™ Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab's Neutrodine™ Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured floor-mount fume hood that requires NO DUCTING for easy installation! Neutrodine Unisorb filters are compatible with over 700 chemicals.

Features (All Models)

  • Eco-Foil™ air foil with aerodynamic Clean-Sweep™ airflow openings
  • Cord-Keeper™ Slots on left and right side of air foil
  • Single piece, tempered safety glass vertical-rising sashes with cable pulley, sash weight and powder-coated aluminum sash handle and tracks with Clean-Sweep™ airflow openings. Upper sash stop at 16 in. sash open height. Upper sash opens to 26 in. high for loading chemicals. Lower sash opens up from floor to 55.2 in. for transporting carts, drums and apparatus
  • Removable front and side panels, and front and interior service access panels for access to plumbing and electrical wiring
  • eGuard2 software communication
  • Smart control module with wifi, Bluetooth and ethernet connectivity
  • Factory-installed command module, fan button and light button
  • Rear baffle
  • By-pass airflow design
  • Glacier white powder-coated steel exterior
  • Chemical-resistant, fiberglass-reinforced, composite panel liner
  • Pre-wired vapor-proof LED lights for up to 63 foot candles rated for 50,000 hours: 2 lights per 4 ft. model, 3 lights per 5 ft. model, 4 lights per 6 ft. model, and 5 lights per 8 ft. model
  • Factory-prepared for up to two electrical duplex receptacles and six service fixtures
  • Roughing prefilter: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model
  • Built-in exhaust fans: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model, to maintain 60 fpm face velocity with only an upper sash opening height of 16 in.
  • Sound pressure of <60 dB(A) with sash at operating height and <48 dB(A) with sash closed
  • Sensor package for primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position, and temperature
  • Audible/visual alarms for breakthrough detection, temperature ≥40°C (104°F), fan failure and high sash position opening. At ≥60°C (140°F), all fans stop
  • Power cord with plug

Fixtured Models May Feature

  • Two pre-plumbed service fixtures with forged brass valves, lower right side with brass tubing for gas and lower left side with copper tubing for cold water. Components for converting either or both fixtures to air and vacuum are provided. Inlet tubing is not provided
  • One pre-wired GFCI electrical duplex receptacle on lower right side

Required Accessories (Not Included)

  • Neutrodine™ Unisorb Filters (except HEPA only models which require HEPA filters). Number of filters required is determined by the width of the hood

Options Include

  • HEPA filter
  • Region-specific electrical plug configurations
  • Service fixtures

Standards Conformance and Regulations

  • SEFA 9, DHIII
  • SEFA 8, Cabinet Surface Finish Tests
  • MODIFIED ASHRAE 110
  • ANSI Z9.5-2011
  • AFNOR NF-X 15-211
  • ASTM E84-09C
  • UL Standard 61010-1
  • CAN/CSA C22.2 No. 61010-1
Specifications

Specifications

UL 61010-1; CAN/CSA C22.2 No. 61010.1; ANSI Z9.5; SEFA 9, DH III; Modified ASHRAE 110; ASTM E84-09 C; AFNOR NF-X 15-211; SEFA 8, Cabinet Surface Finish Tests
General Chemistry; Large Equipment
102.2 in.
LED rated for 50,000 hours
Floor-Mounted
Protector™ EchoTM Floor-Mounted
Filtered Fume Hood
Glacier White
37.7 in.
50/60 Hz
Chemical-resistant, fiberglass-reinforced, composite panel
Powder-Coated Steel
52 in.
5 ft.

Filters required (sold separately)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit