Learn More
Labconco™ Protector™ Echo™ Floor-Mounted Filtered Fume Hoods: 6 ft. Width
Fully-featured, energy-efficient fume hood provides an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.
$53055.00 - $56375.00
Specifications
Product Type | Filtered Fume Hood |
---|---|
Coating | Powder |
Lighting Type | LED |
Product Line | Protector™ Echo™ Floor-Mounted |
Depth (Metric) | 141.5 cm |
Alert:
The Lifetime Payback Questionnaire (LPQ) form is REQUIRED for all Protector Echo Floor-Mounted Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.
Requires: Filters, Remote Blower, Ductwork
Catalog Number | Mfr. No. | Depth (English) | Includes | Filter Type | Amperage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (English) | Includes | Filter Type | Amperage | Price | Quantity & Availability | ||
13-118-800
![]() |
Labconco™
183637200 |
55.7 in. | 0 fixtures, 0 electrical receptacles | Carbon |
Each for $53,055.00
|
|
|||
13-118-801
![]() |
Labconco™
183637201 |
55.7 in. | 2 fixtures, 0 electrical receptacles | Carbon |
Each for $54,240.00
|
|
|||
13-118-802
![]() |
Labconco™
183637202 |
55.7 in. | 2 fixtures, 1 electrical receptacle | Carbon |
Each for $54,400.00
|
|
|||
13-118-803
![]() |
Labconco™
183637220 |
46.7 in. | 0 fixtures, 0 electrical receptacles | Carbon |
Each for $53,055.00
|
|
|||
13-118-804
![]() |
Labconco™
183637221 |
55.7 in. | 2 fixtures, 0 electrical receptacles | Carbon |
Each for $54,240.00
|
|
|||
13-118-805
![]() |
Labconco™
183637400 |
46.7 in. | 0 fixtures, 0 electrical receptacles | Carbon with Acid Sensor |
Each for $55,030.00
|
|
|||
13-118-806
![]() |
Labconco™
183637401 |
46.7 in. | 2 fixtures, 0 electrical receptacles | Carbon with Acid Sensor |
Each for $56,215.00
|
|
|||
13-118-807
![]() |
Labconco™
183637402 |
46.7 in. | 2 fixtures, 1 electrical receptacle | Carbon with Acid Sensor |
Each for $56,375.00
|
|
|||
13-118-808
![]() |
Labconco™
183637420 |
46.7 in. | 0 fixtures, 0 electrical receptacles | Carbon with Acid Sensor |
Each for $55,030.00
|
|
|||
13-118-809
![]() |
Labconco™
183637421 |
46.7 in. | 2 fixtures, 0 electrical receptacles | Carbon with Acid Sensor |
Each for $56,215.00
|
|
|||
13-118-810
![]() |
Labconco™
183637600 |
46.7 in. | 0 fixtures, 0 electrical receptacles | Carbon with Formaldehyde Sensor |
Each for $55,030.00
|
|
|||
13-118-811
![]() |
Labconco™
183637601 |
46.7 in. | 2 fixtures, 0 electrical receptacles | Carbon with Formaldehyde Sensor |
Each for $56,215.00
|
|
|||
13-118-812
![]() |
Labconco™
183637602 |
55.7 in. | 2 fixtures, 1 electrical receptacle | Carbon with Formaldehyde Sensor |
Each for $56,375.00
|
|
|||
13-118-813
![]() |
Labconco™
183637620 |
46.7 in. | 0 fixtures, 0 electrical receptacles | Carbon with Formaldehyde Sensor |
Each for $55,030.00
|
|
|||
13-118-814
![]() |
Labconco™
183637621 |
55.7 in. | 2 fixtures, 0 electrical receptacles | Carbon with Formaldehyde Sensor |
Each for $56,215.00
|
|
|||
13-118-815
![]() |
Labconco™
183637800 |
46.7 in. | 0 fixtures, 0 electrical receptacles | HEPA |
Each for $53,055.00
|
|
|||
13-118-816
![]() |
Labconco™
183637801 |
55.7 in. | 2 fixtures, 0 electrical receptacles | HEPA |
Each for $54,240.00
|
|
|||
13-118-817
![]() |
Labconco™
183637802 |
46.7 in. | 2 fixtures, 1 electrical receptacle | HEPA |
Each for $54,400.00
|
|
|||
13-118-818
![]() |
Labconco™
183637820 |
46.7 in. | 0 fixtures, 0 electrical receptacles | HEPA |
Each for $53,055.00
|
|
|||
13-118-819
![]() |
Labconco™
183637821 |
55.7 in. | 2 fixtures, 0 electrical receptacles | HEPA |
Each for $54,240.00
|
|
|||
Description
The Protector™ Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab's Neutrodine™ Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured floor-mount fume hood that requires NO DUCTING for easy installation! Neutrodine Unisorb filters are compatible with over 700 chemicals.
Features (All Models)
- Eco-Foil™ air foil with aerodynamic Clean-Sweep™ airflow openings
- Cord-Keeper™ Slots on left and right side of air foil
- Single piece, tempered safety glass vertical-rising sashes with cable pulley, sash weight and powder-coated aluminum sash handle and tracks with Clean-Sweep™ airflow openings. Upper sash stop at 16 in. sash open height. Upper sash opens to 26 in. high for loading chemicals. Lower sash opens up from floor to 55.2 in. for transporting carts, drums and apparatus
- Removable front and side panels, and front and interior service access panels for access to plumbing and electrical wiring
- eGuard2 software communication
- Smart control module with wifi, Bluetooth and ethernet connectivity
- Factory-installed command module, fan button and light button
- Rear baffle
- By-pass airflow design
- Glacier white powder-coated steel exterior
- Chemical-resistant, fiberglass-reinforced, composite panel liner
- Pre-wired vapor-proof LED lights for up to 63 foot candles rated for 50,000 hours: 2 lights per 4 ft. model, 3 lights per 5 ft. model, 4 lights per 6 ft. model, and 5 lights per 8 ft. model
- Factory-prepared for up to two electrical duplex receptacles and six service fixtures
- Roughing prefilter: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model
- Built-in exhaust fans: 2 each per 4 ft. model, 3 each per 5 ft. model, 4 each per 6 ft. model, and 5 each per 8 ft. model, to maintain 60 fpm face velocity with only an upper sash opening height of 16 in.
- Sound pressure of <60 dB(A) with sash at operating height and <48 dB(A) with sash closed
- Sensor package for primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position, and temperature
- Audible/visual alarms for breakthrough detection, temperature ≥40°C (104°F), fan failure and high sash position opening. At ≥60°C (140°F), all fans stop
- Power cord with plug
Fixtured Models May Feature
- Two pre-plumbed service fixtures with forged brass valves, lower right side with brass tubing for gas and lower left side with copper tubing for cold water. Components for converting either or both fixtures to air and vacuum are provided. Inlet tubing is not provided
- One pre-wired GFCI electrical duplex receptacle on lower right side
Required Accessories (Not Included)
- Neutrodine™ Unisorb Filters (except HEPA only models which require HEPA filters). Number of filters required is determined by the width of the hood
Options Include
- HEPA filter
- Region-specific electrical plug configurations
- Service fixtures
Standards Conformance and Regulations
- SEFA 9, DHIII
- SEFA 8, Cabinet Surface Finish Tests
- MODIFIED ASHRAE 110
- ANSI Z9.5-2011
- AFNOR NF-X 15-211
- ASTM E84-09C
- UL Standard 61010-1
- CAN/CSA C22.2 No. 61010-1
Specifications
Filtered Fume Hood | |
LED | |
141.5 cm | |
General Chemistry; Large Equipment | |
52 in. | |
6 ft. |
Powder | |
Protector™ Echo™ Floor-Mounted | |
118.6 cm | |
102.2 in. | |
259.6 cm | |
182.9 cm |
Filters required (sold separately)
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.