Learn More
Labconco™ Protector™ Filtered Fume Hood Accessories
SDP
Filters and accessories for use with Protector Echo and Airo Filtered Fume Hoods.
Supplier:  Labconco™ 9577421
 View more versions of this product
Catalog No. 13-118-001
Add to Cart
Description
Description
  • These accessories are for use with Labconco Protector Echo and Airo Filtered Fume Hoods
  • For all filters and prefilters: the number of filters/prefilters required depends on the width of the hood and if it is an initial order or a replacement order
Neutrodine Unisorb molecular carbon filter for multi-purpose use with acids/bases/solvents, separated or combined. Compatible with 700+ chemicals.HEPA Filter, 99.995% efficient, H14 type.Replacement roughing prefilter, located in light box.Replacement acid roughing prefilter, located in light box. Treated for use with acids.Replacement ammonia roughing prefilter, located in light box. Treated for use with ammonia.Acid Gas sensor is required for proper hood function on models with acid sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).Formaldehyde Gas Sensor is required for proper hood function on models with formaldehyde sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).
Specifications
Specifications
0.99995
Protector Echo and Airo Filtered Fume Hoods
11 lb.
30 in.
16 in.
76.2 cm
40.6 cm
HEPA
4 in.
5.0 Kg
30 in.
10.2 cm
76.2 cm
HEPA Filter
Product Suggestions
Product Suggestions