Learn More
Labconco™ Protector™ Filtered Fume Hood Accessory, RFID Administrator Authorization Card
SDPSCI_ED
For use with Protector Echo™ and Airo™ Filtered Fume Hood. Labconco Protector Filtered Fume Hood Accessory. authorization cards are required for use and maintenance providing additional safety and data/usage records.
Supplier:  Labconco™ 9580900
Catalog No. S28330
Explore available promotions
Add to Cart
Description
Description
  • Radio Frequency Identification Administration Card
  • Provides full access to view stored information on fume hood
  • Required to operate/view stored data on fume hood, including user time usage
Warranty and Services
One year
Specifications
Specifications
RFID Administrator Card
5.39 cm
1 lb.
3.375 in.
2.125 in.
Protector Echo and Airo Filtered Fume Hood
453.5 g
8.57 cm
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov