Learn More
UbpbioSupplier Diversity Partner PROTEIN A AGAROSE, 1ML DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner P50201
Protein Purification Reagents – Protein A Resin (UBPBio catalog# P5020-1, P5020-5, P5020-25) – Protein A XPure Agarose Resin consists of recombinant Protein A covalently bound to crosslinked agarose beads, which greatly minimizes leakage of Protein A and allow reuse the affinity resin. The resin with a binding capacity of ~ 20 mg human IgG/ml resin and works well in batch or FPLC column purifications.
Catalog No. 50-112-6957