Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Protein A - Alkaline Phosphatase

Protein A is 42 kDa surface protein of bacterium Staphylococcus aureus, which binds Fc region of many mammalian IgGs.

$180.10 - $351.00

Specifications

Product Type Source: E. coli
Storage Requirements Store at -20° C.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
20-010-100 View Documents Novus Biologicals™
20010100
0.1mg Each for $180.10
Add to cart
 
20-011-000 View Documents Novus Biologicals™
20011000
1mg Each for $351.00
Add to cart
 
Description

Description

Protein A is a 42kDa surface protein of the bacterium Staphylococcus aureus, which binds the Fc region of many mammalian IgGs. Its high affinity for immunoglobulins is the reason that it is often used in biochemical research. Alkaline Phosphatase is a hydrolase enzyme. It is extracted from bovine intestine and purified using multiple precipitation, fractionation, chromatography, and filtration steps. Alkaline Phosphatase is frequently employed in immunoassays, and a commonly used colorimetric substrate for it is PNPP; the product of this reaction, p-nitrophenol, can be read at 405nm. Fluorogenic substrates such as 4- methylumbelliferyl phosphate (MUP) may also be used.
Specifications

Specifications

Source: E. coli
Store at -20° C.

For research use only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit