GE Healthcare Protein A and Protein G Mag Sepharose™ Promotion

Manufacturer: GE Healthcare 28951379PM

 View more versions of this product

Catalog No. 45-002-173PM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Application) Designed for covalent coupling of any Protein or biomolecule containing a free Amine group and is used for various types of pull down experiments, for biomarker research
    Ligand Protein G
    Quantity 4 x 500μL
    Binding Capacity 13 to 22mg Human IgG/mL
    Storage Requirements 4° to 8°C
    Matrix Paramagnetic spherical, highly cross linked agarose particles
    Particle Size 37 to 100μm
    Bead Type Protein G Mag Sepharose

    Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

    There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

    Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

    • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
    • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
    • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
    • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

    Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
    Ligand: Native protein A; Native protein G
    Binding capacity Protein A: 8-17mg human IgG/mL gel
    Binding capacity Protein G: 13-22mg human IgG/mL gel
    Particle size: 37-100µm
    Working temperature: Room temperature
    Storage solution: 20% ethanol
    Storage temperature: 4°C to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit