Learn More
Aat BioquestSupplier Diversity Partner ReadiLink™ Protein Biotinylation Kit DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5520
Biotin is widely used for labeling biomolecules, in particular, antibodies. Our ReadiLink™ Protein Biotinylation Kit is optimized for the preparation of biotin-labeled IgG for enzyme immunoassay (EIA).
Catalog No. 50-195-3288