Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA586875
Description
Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55kDa and the normal concentrations in human plasma is approximately 1-3 ng/ml because of the very fast turnover, the proenzyme concentration is approximately 3 μg/ml. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulmis, and recurrent venous thrombosis.Specifications
Protein C | |
Polyclonal | |
Unconjugated | |
PROC | |
Vitamin K-dependent protein C (EC:3.4.21.69); Anticoagulant protein C; Autoprothrombin IIA; Blood coagulation factor XIV; Vitamin K-dependent protein C light chain; Vitamin K-dependent protein C heavy chain; Activation peptide; PROC | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19123, 25268, 5624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04070, P31394, P33587 | |
Proc | |
A synthetic peptide corresponding to residues in Human PROC. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title