Learn More
BioVision IncSupplier Diversity Partner Protein Carbonyl Content Assay Kit: Protein Carbonyl Content Assay Kit: Simple and Accurate Colorimetric Assay for Quantifying carbonyls in protein samples. Detection Limit: 0.15 nmol/mg. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K830100
Protein carbonyl groups are an important and immediate biomarker of oxidative stress. DNPH tagging of protein carbonyls has been one of the most common measures of oxidative stress. DNP hydrazones formed from the reaction are easily quantifiable at 375 nm. BioVision’s Protein Carbonyl Content Assay Kit is designed to provide a simple and accurate method of quantifying carbonyls in protein samples. Using BSA as an example, a 1 mg (~ 15 nmol) sample has a detection limit of about 0.15 nmol carbonyl, where BSA typically contains approximately 1-3 nmol carbonyl/mg. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9944