Learn More
Cell Biolabs IncSupplier Diversity Partner PROTEIN CARBONYL ELISA TRL SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA310T
The OxiSelect Protein Carbonyl ELISA Kit (Trial Size) quantifies protein carbonyl content, a stable marker of oxidative stress, in plasma, serum, cell lysates or purified proteins. 32 wells including standards and unkwown samples.
Catalog No. 50-673-494