Learn More
Protein Disulfide Isomerase/P4HB Mouse anti-Human, Clone: 3G5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP17918520UL
 View more versions of this product
Catalog No. NBP17918520
Add to cart
Description
Description
Protein Disulfide Isomerase/P4HB Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications
Specifications
Protein Disulfide Isomerase/P4HB
3G5
Western Blot 2.5 ug/ml
NP_000909
Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55
Mouse
IgM Kappa
Protein A or G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
P4HB
The immunogen for this antibody is P4HB.
58 kDa
20ul
Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia
Primary
5034