Learn More
BioworldSupplier Diversity Partner Protein Extraction Wash Buffer, 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400482
Protein Extraction Wash Buffer Sample buffer for protein extraction and purification of GST-fusion proteins. Technical Information: Tris-HCl, pH 8.0 (50mM)NaCl (500mM)Triton X 100 (1%)
Catalog No. 50-253-049