Learn More
BioworldSupplier Diversity Partner PROTEIN G (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220606783
Recombinant Protein G Recombinant Protein G contains only two IgG binding domain, but lacks the albumin binding domain to ensure the maximum specific IgG binding capacity. Protein G binds to all IgG subclasses from human, mouse and rat ...
Catalog No. 50-253-778