Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFN-alpha 11 Protein

A New rmIFN-alpha11 is Available! It is mammalian cell derived, has lower endotoxin specifications, is made in-house, and is priced competitively!

Manufacturer:  R&D Systems™ 121251

Catalog No. 12-125-1R


Add to cart
This item is not returnable. View return policy

Description

Description

  • Alternate Names: IFNA11
Specifications

Specifications

IFN-alpha 11
100000 UN
Ifna11
Unconjugated
15964
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant Mouse IFN-alpha 11 Protein
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit