Learn More
Alfa Aesar™ Protein Kinase C (19-36)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66426MCR
 View more versions of this product
Catalog No. AAJ66426MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2151.51
1mg
Arg-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-Lys-Asn-Val-His-Glu-Val-Lys-Asn
-30° to -10°C
Protein Kinase C (19-36)
HPLC
Solid
RUO
PKC Inhibitor Peptide