Learn More
Alfa Aesar™ Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66521EXD
 View more versions of this product
Catalog No. AAJ66521EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
843.98
0.5mg
Solid
RUO
Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
HPLC
A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val
-30° to -10°C