Learn More
Eppendorf™ Protein LoBind Tubes
Manufacturer:  Eppendorf™ 2231000342
 View more versions of this product
Catalog No. 05-412-693
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Features:
  • Eppendorf LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing to minimize evaporation
Specifications
Specifications
Preparation or storage of protein, peptide or antibody samples; Enzymatic assays; Storage of virus stok solutions and cell suspensions
100 (2 bags x 50)
PCR clean
Centrifuge
5.0mL