Learn More
Eppendorf™ Protein LoBind Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Eppendorf™ 2231000348
 View more versions of this product
Catalog No. 05-412-699
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Features:
  • Eppendorf LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing to minimize evaporation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.