Learn More
GE Healthcare Protein and Peptide Molecular Weight Markers: PROMO
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. GE Healthcare Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
Manufacturer:  GE Healthcare 17044501PM
 View more versions of this product
Catalog No. 45-000-071LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Get clear, sharp bands for easy measurement of migration distances
  • Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
Specifications
10 Vials
Solid
4°C
MW determinations by Polyacrylamide Gel electrophoresis (PAGE) in homogeneous or gradient gels
Thyroglobulin: 76μg, 669000 (Mr), porcine thyroid (source), Ferritin: 50μg, 440000 (Mr), equine spleen (source), Catalase: 36μg, 232000 (Mr), bovine liver (source), Lactate dehydrogenase
Protein and Peptide Molecular Weight Markers