Learn More
GE Healthcare Protein and Peptide Molecular Weight Markers: PROMO
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. GE Healthcare Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
Specifications
Quantity 10 Vials
Storage Requirements 4°C
View More Specs
Products 2
Catalog Number Mfr. No. Kit Contents Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Kit Contents Product Type Price Quantity & Availability  
45-000-071LR View Documents GE Healthcare
17044501PM
Thyroglobulin: 76μg, 669000 (Mr), porcine thyroid (source), Ferritin: 50μg, 440000 (Mr), equine spleen (source), Catalase: 36μg, 232000 (Mr), bovine liver (source), Lactate dehydrogenase Protein and Peptide Molecular Weight Markers This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-000-072LR View Documents GE Healthcare
17044601PM
Phosphorylase b: 67μg, 97000 (Mr), Rabbit muscle (source), Albumin: 83μg, 66000 (Mr), Bovine Serum (source), Ovalbumin: 147μg, 45000 (Mr), Chicken Egg white (source), Carbonic Anhydrase Protein and Peptide Molecular Weight Markers This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Get clear, sharp bands for easy measurement of migration distances
  • Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
Specifications
10 Vials
4°C