Learn More
BioworldSupplier Diversity Partner Protein Pre-Cleaning Buffer, 20 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400491
Protein Precleaning Buffer, 2 X 10ml For clearification of proteins to eliminate nucleic acids and polysaccharides before electrophoresis. Total protein preparation and cleaning solution Technical Information:For use with 1D gels Storage: 4°C Sold as 2 - 10 ml quantities of ready-to-use solution each
Catalog No. 50-253-050