Learn More
BioworldSupplier Diversity Partner Protein pur-E.coli Elution Buffer, 250 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400501
Protein Purification Buffer, E.coli Lysis Sample buffer for HIS-TAG protein purification from E.coli and insect cells. Technical Information: pH 8.0 Na2HPO4 (50mM)NaCl (300mM)Imidazol (250mM)
Catalog No. 50-253-051