Learn More
BioworldSupplier Diversity Partner Protein pur-E.coli wash Buffer, 250 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400521
Protein Purification Buffer, E.coli Lysis Sample buffer for HIS-TAG protein purification from E.coli and insect cells. Technical Information: pH 8.0 Na2HPO4 (50mM) NaCl (300mM) Imidazol (10mM)
Catalog No. 50-253-055