Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein S/PROS1 Mouse anti-Human, Clone: 8D5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105150
Description
Protein S/PROS1 Monoclonal specifically detects Protein S/PROS1 in Human samples. It is validated for ELISA, Western Blot (Negative).Specifications
Protein S/PROS1 | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000, Western Blot (Negative) | |
EC 3.4.21, PROS, protein S (alpha), protein Sa, PS21, PS22, PS23, PS24, PS25, PSA, vitamin K-dependent plasma protein S, vitamin K-dependent protein S | |
Mouse | |
75 kDa | |
100 μg | |
Primary | |
Protein S | |
Store at 4C in the dark. | |
IgG2b κ |
ELISA, Western Blot | |
8D5 | |
Unconjugated | |
P07225 | |
PROS1 | |
Protein S isolated from human plasma adsorbed onto aluminum hydroxide gel. | |
Protein A or G purified | |
RUO | |
5627 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction