Learn More
BioworldSupplier Diversity Partner Protein Solubilization Buffer, 50 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220200012
Protein Solubilization Buffer.Rehydration Buffer for Total Protein Solubilization from acetone Powder.Contains: 7M Urea, 2% CHAPS, 2M Thiourea, 0.2% DTT
Catalog No. 50-252-981