Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteinase K, Ready-to-Use Soln.

A stable and highly reactive serine protease; used for protein and nucleic acid isolation

$233.00 - $891.00

Specifications

CAS 39450-01-6
Formula Weight 28.9 kd
Storage Requirements −30° to −10°C
Format Liquid
For Use With (Application) A stable and highly reactive serine protease; used for protein and nucleic acid isolation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ63710AC View Documents Alfa Aesar™
J63710AC
25mL Each for $891.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ63710AA View Documents Alfa Aesar™
J63710AA
5mL Each for $233.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

39450-01-6
−30° to −10°C
A stable and highly reactive serine protease; used for protein and nucleic acid isolation
DNA Extraction Reagent
28.9 kd
Liquid
Clear, Colorless
Safety and Handling

Safety and Handling

EINECSNumber : 254-457-8

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit