Learn More
MilliporeSigma™ Proteinase K Solution, 600mAU/mL
Efficient removal of proteins from nucleic acid solutions
$128.00 - $388.00
Description
- Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album
- The enzyme exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA
- Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50-60°C)
- The recommended working concentration is 50-100μg/mL for protein removal and enzyme inactivation, and up to 2mg/mL for tissue treatment
- The Proteinase K, Lyophilized powder can be prepared as a 20mg/mL stock solution in water and stored in aliquots at -20°C
- The enzyme is also available as a ready-to-use concentrated stock solution (600mAU/mL) that is convenient for routine use in most applications
- 1mg of Proteinase K is the equivalent of 30mAU (AU = Anson unit)
- The Novagen Proteinase K products are free of detectable DNase and RNase
Specifications
-20°C | |
Proteinase K |