Learn More
Proteinase K, Tritirachium album limber, Alfa Aesar™
Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.
Manufacturer:  Alfa Aesar J62051MC
 View more versions of this product
Catalog No. AAJ62051MC
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Lyophilized Powder, 28.9 kDa
100mg