Learn More
Proteinase K, Tritirachium album limber, Alfa Aesar™
Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.
Manufacturer:  Alfa Aesar J62051MB
 View more versions of this product
Catalog No. AAJ62051MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Lyophilized Powder, 28.9 kDa
25mg
Lyophilized powder,28.9 kDa
Safety and Handling
Safety and Handling
EINECSNumber : 254-457-8
Recommended Storage : Keep cold