Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteinase K, Tritirachium album limber, Alfa Aesar™

Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.

$54.40 - $114.80

Specifications

Format Lyophilized Powder, 28.9 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62051MB View Documents Alfa Aesar
J62051MB
25mg Each for $54.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ62051MC View Documents Alfa Aesar
J62051MC
100mg Each for $114.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Lyophilized Powder, 28.9 kDa
Safety and Handling

Safety and Handling

EINECSNumber : 254-457-8

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit