Learn More
MilliporeSigma™ ProteoExtract™ Albumin/IgG Removal Kit, Maxi
Specialized extraction kit with high capacity, reusable columns specifically designed for rapid and efficient removal of albumin and IgG from serum body fluid samples
Supplier:  MilliporeSigma™ 1226431KIT
Catalog No. 12-264-31KIT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • High capacity, reusable columns specifically designed for rapid and efficient removal of >80% of albumin (column 1 containing albumin-affinity resin) and IgG (column 2 containing immobilized protein A) from body fluid samples, either on an LC-instrument or manually, attached to a syringe
  • Removal of high abundance proteins allows enhanced detection of low abundance proteins
Specifications
Specifications
Store at+2°C to+8°C
Affinity column
20 Albumin-containing samples (60-180μL each) and 10 IgG-containing samples (60-180μL each)
1 kit
Two Albumin removal columns, one IgG Removal Column, 10X Albumin/IgG binding buffer, Albumin elution buffer, IgG Elution Buffer, and user protocol
Product Suggestions
Product Suggestions