Learn More
MilliporeSigma™ ProteoExtract™ Albumin/IgG Removal Kit
Specialized extraction kit with columns specifically designed for rapid and efficient removal of albumin/IgG from serum body fluid samples
Manufacturer:  MilliporeSigma™ 1226421KIT
Catalog No. 12-264-21KIT
Add to cart
Description
Description
  • Columns are specifically designed for rapid and efficient removal of >80% of albumin and IgG from serum body fluid samples
  • Removal of high abundance proteins allows for enhanced detection of low abundance proteins
Specifications
Specifications
1 kit
12 disposable columns packed with Albumin/IgG-Removal Resin, binding buffer, and a user protocol
12 plasma samples (20-60μL each)
Affinity column
Store at+2°C to+8°C