Learn More
MilliporeSigma™ ProteoExtract™ Albumin Removal Kit, Maxi
Specialized extraction kit with high capacity, reusable columns specifically designed for rapid and efficient removal of albumin from serum body fluid samples
Supplier: MilliporeSigma™ 1226411KIT
Description
- High capacity, reusable columns specifically designed for the rapid and efficient removal of >80% of albumin from body fluid samples, either on an LC-instrument or manually (attached to a syringe)
- Removal of these high abundance proteins allows enhanced detection of low abundance proteins
- Each column can be re-used up to 10 times
- When used manually, the albumin removal columns are operated as syringe-tip filters
- when used with LC-instrument control, the columns can be connected to any standard HPLC/FPLC instrument via a 10/32 in. port using the provided adapters
Specifications
Store at+2°C to+8°C | |
Affinity column | |
20 Samples (60-180μL each) |
1 kit | |
Two Albumin removal columns, 10X Albumin/IgG binding buffer, Albumin elution buffer, and user protocol |