Learn More
MilliporeSigma™ ProteoExtract™ Albumin Removal Kit Available on GSA/VA Contract for Federal Government customers only.
Specialized extraction kit with columns specifically designed for rapid and efficient removal of albumin from serum body fluid samples
Manufacturer:  MilliporeSigma™ 1226401KIT
Catalog No. 12-264-01KIT
Add to cart
Description
Description
  • Columns are specifically designed for rapid and efficient removal of >80% of albumin from serum body fluid samples
  • Removal of albumin allows for enhanced detection of low abundance proteins
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1 kit
12 disposable columns packed with albumin-removal resin, binding buffer, and user protocol
12 plasma samples (20-60μL each)
Affinity column
Store at+2°C to+8°C