Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expedeon Proteoloc™ Protease Inhibitor Cocktail EDTA-Free

Protects protein extracts from aminopeptidases, metalloproteases, and serine, cysteine, and aspartic acid proteases.

$80.00 - $619.00

Specifications

Product Type Proteoloc™ Protease Inhibitor Cocktail EDTA-Free
Composition AEBSF, Aprotinin, Bestatin, E-64, Leupeptin, Pepstatin A
Concentration 100X stock solutions in DMSO
For Use With (Application) Technique which are incompatible with chelation
Format Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EX42517 View Documents Expedeon
42517
3 x 10mL Each for $619.00
Add to cart
 
EX42516 View Documents Expedeon
42516
3mL Each for $80.00
Add to cart
 
Description

Description

  • Used with isoelectric focusing, metal-affinity column separation, or another technique that is incompatible with chelation
  • General-purpose reagent suitable for protease inhibition during protein extraction
  • Provided as 100X stock solutions in DMSO
Specifications

Specifications

Proteoloc™ Protease Inhibitor Cocktail EDTA-Free
100X stock solutions in DMSO
Liquid
AEBSF, Aprotinin, Bestatin, E-64, Leupeptin, Pepstatin A
Technique which are incompatible with chelation
Store at 4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit