Learn More
Sepax Technologies Inc (433NP5-4610) HPLC Column, Sepax, Proteomix HIC Phenyl-NP5, 5um, NP 4.6 x 100 mm
Supplier:  Sepax Technologies Inc 433NP54610
Encompass_Preferred
Analytical and Preparative Hydrophobic Interaction Chromatography from Sepax Technologies, Inc. || Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica-based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with a wide pH range (2-12) and high chemical stability. The nonporous structure, unique chemistry, and narrow particle distribution offer special selectivity, high-resolution separation of proteins such as mAb, ADC and related protein fragments, DNA and oligonucleotides.Sepax also provides polymethacrylate-based Generik MC and Polar MC HIC 30 m media which offers similar characteristics to the Proteomix HIC-NP but is designed for process-scale applications from grams to kilograms of proteins. For full product details, additional dimensions, rush orders, and custom requests, please contact the manufacturer.
Catalog No. NC1141460