Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Proteomix™ HIC Phenyl-NP5 Column, 5 μm Particle Size

Proteomix™ HIC columns are specially designed for high resolution and high efficiency separations of proteins, oligonucleotides and peptides.

$1286.00 - $1547.00

Specifications

Material Stainless Steel Hardware
Matrix Phenyl Phase/Polystyrene/Divinylbenzene Particle Platform (Spherical, highly-crossed linked)
Particle Size 5 μm
Phase Hydrophobic Interaction
Pore Size Nonporous
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
11-100-6520
View Documents
MilliporeSigma™ Supelco™
61874U
4.6 mm 35 mm
Each for $1,286.00
Add to cart
 
11-100-6390
View Documents
MilliporeSigma™ Supelco™
61878U
7.8 mm 50 mm
Each for $1,547.00
Add to cart
 
11-100-6389
View Documents
MilliporeSigma™ Supelco™
61876U
4.6 mm 100 mm
Each for $1,547.00
Add to cart
 
Description

Description

Utilizing proprietary surface technologies, Proteomix HIC-NP resin is made of non-porous polystyrenedivinylbenzene (PS/DVB) beads with narrow-dispersed particle size distribution. The PS/DVB bead is modified with alkyl groups or aryl group that provides hydrophobic interaction with analytes. Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with wide pH range (2-12) and high thermal stability. The nonporous structure and narrow particle distribution offer special selectivity, high resolution separation of proteins such as mAb (monoclonal antibody), ADC (antibody drug conjugate) and related protein fragments, DNA and oligonucleotides. Proteomix HIC-NP media is applicable at laboratory discovery, laboratory-scale purification and process chromatography for the production of a few mgs to kilogram of proteins.
Specifications

Specifications

Stainless Steel Hardware
5 μm
Nonporous
Phenyl
Phenyl Phase/Polystyrene/Divinylbenzene Particle Platform (Spherical, highly-crossed linked)
Hydrophobic Interaction
Proteomix
HIC HPLC
SDS
Product Certifications

Proteomix is a registered trademark of Sepax Technologies

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit